Learn More
Chemglass Life Sciences 100mL Single Neck Evaporating Flask, 24/40 Outer Joint, Plastic Coated Recovery Flask
SDP
Catalog No. CG1512P03
Encompass_Preferred
100mL Single Neck Evaporating Flask, 24/40 Outer Joint, Plastic Coated Recovery Flask. Similar to CG-1512 recovery flasks, but plastic coated to prevent injury from broken glass and spillage of hazardous materials.
Catalog No. CG1512P03 Supplier Chemglass Life Sciences Supplier No. CG1512P03
100mL Single Neck Evaporating Flask, 24/40 Outer Joint, Plastic Coated Recovery Flask. Similar to CG-1512 recovery flasks, but plastic coated to prevent injury from broken glass and spillage of hazardous materials.