Learn More
Chemglass Life Sciences 20L Flask, Evaporating, Large Scale, 145mm Flange, 440mm OAH
SDP
Catalog No. 501531087
Encompass_Preferred
20L Flask, Evaporating, Large Scale, 145mm Flange, 440mm OAH. Flasks are compatible with Buchi large scale rotary evaporators. Flasks to fit large scale rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation.
Catalog No. 50-153-1087 Supplier Chemglass Life Sciences Supplier No. CG133714
20L Flask, Evaporating, Large Scale, 145mm Flange, 440mm OAH. Flasks are compatible with Buchi large scale rotary evaporators. Flasks to fit large scale rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation.