Learn More
Ace Glass, Inc. Flask, Fernbach, 1800mL, baffled, 38mm OD plain neck, cs/3
SDP
Catalog No. 50971395
Encompass_Preferred
1800mL baffled Fernbach culture flask with 38mm O.D. long style straight neck for either plastic or stainless steel closures. The bottom of the flask is triple baffled to achieve maximum oxygen transfer to the culture medium. CAUTION: Do not place on direct heat source.
Catalog No. 50-971-395 Supplier Ace Glass, Inc. Supplier No. 387520
1800mL baffled Fernbach culture flask with 38mm O.D. long style straight neck for either plastic or stainless steel closures. The bottom of the flask is triple baffled to achieve maximum oxygen transfer to the culture medium. CAUTION: Do not place on direct heat source.