Learn More
Ace Glass, Inc. Flask, Fernbach, 2800mL, triple baffled, #13 stopper size beaded neck, cs/3
SDP
Catalog No. 50971394
Encompass_Preferred
Flask, Fernbach, 2800mL, triple baffled, #13 stopper size beaded neck, cs/3
Catalog No. 50-971-394 Supplier Ace Glass, Inc. Supplier No. 387430
Flask, Fernbach, 2800mL, triple baffled, #13 stopper size beaded neck, cs/3