Learn More
Ace Glass, Inc. Flask, Fernbach, 2800mL, baffled, GL45 neck with cap
SDP
Catalog No. 50971397
Encompass_Preferred
Fernbach style culture flask with mide-mouth neck and screw cap closure. Supplied with GL45 PP cap. Flask is triple baffled on outside edges for maximum oxygen transfer to the media. Caution! Do not place on direct heat source.
Catalog No. 50-971-397 Supplier Ace Glass, Inc. Supplier No. 387745
Fernbach style culture flask with mide-mouth neck and screw cap closure. Supplied with GL45 PP cap. Flask is triple baffled on outside edges for maximum oxygen transfer to the media. Caution! Do not place on direct heat source.