Learn More
Ace Glass, Inc. Flask, Fernbach, 4000mL, no baffles, 38mm plain neck, cs/1
SDP
Catalog No. 50978206
Encompass_Preferred
4000mL Fernbach style culture flask with 38mm long style straight neck to accomm ODate plastic or stainless steel closures. Flask has a flat bottom, and no baffles. CAUTION: Do not place on direct heat source!
Catalog No. 50-978-206 Supplier Ace Glass, Inc. Supplier No. 387934
4000mL Fernbach style culture flask with 38mm long style straight neck to accomm ODate plastic or stainless steel closures. Flask has a flat bottom, and no baffles. CAUTION: Do not place on direct heat source!