Learn More
Ace Glass, Inc. Flask, filtering, 10L, plastic coated, removable polypropylene hose connection, 82mm OD neck (+/- 5mm), Duran;
SDP
Supplier:  Ace Glass, Inc. 6989121
Encompass
10L heavy wall filtering flask made of Duran glass that is plastic-coated for increased safety. Flask is bottle shaped, with a removable polypropylene hose connection.
Catalog No. 50-973-986