Learn More
Ace Glass, Inc. Flask, filtering, 15L, plain glass, removable polypropylene hose connection, 84mm OD neck(+/- 5mm), Duran;
SDP
Supplier:  Ace Glass, Inc. 698924
Encompass
15L heavy wall filtering flask made of Duran clear glass. Flask is bottle shaped, with a removable polypropylene hose connection.
Catalog No. 50-973-984