Learn More
Wilmad Labglass FLASK FILTER 40/35 1000ML
Supplier:  Wilmad Labglass BP1827000
Encompass_Preferred
Flask, Filter, Heavy Wall, [ST] Joint. Capacity: 1000mL, Joint Size: [ST] 40/35 Inner. Heavy wall flask w/ standard taper 40/35 inner joint on top. Suitable For Reduced Pressuresure filtration.
Catalog No. 50-806-269
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov