Learn More
Ace Glass, Inc. 1L flask, 145mm OD por A disc, 4" flange top, 5/8" OD bottom tube.
SDP
Supplier:  Ace Glass, Inc. 630006
Encompass_Preferred
1L cylindrical, heavy-wall reaction flask with an integral 145mm O.D., 145-174 micron, fritted disc. Flask has a 4 flat flange top with an o-ring groove, and a 5/8 O.D. bottom outlet tube.
Catalog No. 50-972-777