Learn More
Ace Glass, Inc. 6L flask, 177mm OD por A disc, 8" flange top, 1-1/4" tube
SDP
Supplier:  Ace Glass, Inc. 630030
Encompass_Preferred
6L cylindrical, heavy-wall reaction flask with an integral 177mm O.D., 145-174 micron, fritted disc. Flask has a 8 flat flange top with an o-ring groove, and a 1.25 O.D. bottom outlet tube.
Catalog No. 50-972-782