Learn More
Hach Company Flask, Filtering, 1000 mL
Supplier:  Hach Company 54653
Encompass
Glass filtering flask with side-arm at 90° angle. Heavy-walled with graduated capacity scale. Designed to ASTM E1406, Type II, Class I requirements. Hose connection accepts 5/16" I.D. flexible tubing.
Catalog No. 50-199-4373