Learn More
Chemglass Life Sciences Flask, Filtering, 1500mL, Polypropylene, Fits Tubing ID: 3/8", Stopper Size: #10
SDP
Supplier:  Chemglass Life Sciences CG852115L
Encompass_Preferred
Flask, Filtering, 1500mL, Polypropylene, Fits Tubing ID: 3/8", Stopper Size: #10. Heavy-duty filtering flask will withstand vacuum to 30 in.Hg. Molded in one piece polypropylene to withstand full vacuum and for durability. The tubulation is at a 45 angle to prevent tipping when hose is attached. Barbed adapter accepts 0.25 to 0.375 ID tubing. Tubing sold separately.
Catalog No. 50-153-1820