Learn More
Ace Glass, Inc. Flask, flat bottom, 100mL, 24/40
SDP
Catalog No. 50973545
Encompass_Preferred
100mL flat bottom flask with a single, short neck and a 24/40 standard taper outer joint.
Catalog No. 50-973-545 Supplier Ace Glass, Inc. Supplier No. 689521
100mL flat bottom flask with a single, short neck and a 24/40 standard taper outer joint.