Learn More
Ace Glass, Inc. 2L flat bottom flask with a single, short neck and a 24/40 standard taper outer joint.
SDP
Catalog No. 50973550
Encompass_Preferred
2L flat bottom flask with a single, short neck and a 24/40 standard taper outer joint.
Catalog No. 50-973-550 Supplier Ace Glass, Inc. Supplier No. 689528
2L flat bottom flask with a single, short neck and a 24/40 standard taper outer joint.