Learn More
Ace Glass, Inc. Flask, flat bottom, 300mL, 24/40
SDP
Catalog No. 50978922
Encompass_Preferred
300mL flat bottom flask with a single, short neck and a 24/40 standard taper outer joint.
Catalog No. 50-978-922 Supplier Ace Glass, Inc. Supplier No. 689525
300mL flat bottom flask with a single, short neck and a 24/40 standard taper outer joint.