Learn More
Ace Glass, Inc. Flask, flat bottom, 500mL, three neck, 24/40 center, 24/40 sides
SDP
Catalog No. NC9909263
Encompass_Preferred
500mL three neck, flat bottom flask can sit on stirrer hot plate with no further support. Center and angled side necks have 24/40 standard taper joints.
Catalog No. NC9909263 Supplier Ace Glass, Inc. Supplier No. 693915
500mL three neck, flat bottom flask can sit on stirrer hot plate with no further support. Center and angled side necks have 24/40 standard taper joints.