Learn More
Wilmad Labglass FLASK FLTR HW 250ML 24/40 HC
Supplier:  Wilmad Labglass LG7800102
Encompass_Preferred
Filter Flask, Heavy Wall, Erlenmeyer Style, w/ Standard Taper Outer Joint and Side Hose Connection. Capacity: 250mL. Joint Size: [ST] 24/40. Hose Connection:
Catalog No. 50-805-842