Learn More
Ace Glass, Inc. FLASK HEAD ONLY FLASK HEAD ONLY
SDP
Catalog No. 509558128
Encompass_Preferred
Flask head with 29/42 joint, for use with ACE Automatic Dispensing Pipet Apparatus in 1mL to 100mL capacities. Head includes spring and clip.
Catalog No. 50-955-8128 Supplier Ace Glass, Inc. Supplier No. 800410
Add to Cart
Add to Cart
Flask head with 29/42 joint, for use with ACE Automatic Dispensing Pipet Apparatus in 1mL to 100mL capacities. Head includes spring and clip.