Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC0966060
Encompass_Preferred
Aluminum housed heating mantle for 500ml spherical flask to 600 degrees C maximum internal operating temperature, 470W, 115V, CSAThe Series TM mantle covers only the flask's bottom half, letting you see its full contents. Larger sizes have multiple circuits for ease of temperature control when a flask is less than half full. mantles are available with a bottom hole for flask drains.
Catalog No. NC0966060 Supplier Glas Col LLC Supplier No. 100B STM900
Aluminum housed heating mantle for 500ml spherical flask to 600 degrees C maximum internal operating temperature, 470W, 115V, CSAThe Series TM mantle covers only the flask's bottom half, letting you see its full contents. Larger sizes have multiple circuits for ease of temperature control when a flask is less than half full. mantles are available with a bottom hole for flask drains.