Learn More
Andwin Scientific FLASK HEATER 500ML 115VAC-DC
SDP
Catalog No. 11474A
Encompass_Preferred
This 115V aluminum heating mantle is used for 500ml round bottom flask. It can operate up to 650 deg.C. It features rigid housing, fabric interior and 3 in. legs.
Catalog No. 11-474A Supplier Andwin Scientific Supplier No. STM 900
This 115V aluminum heating mantle is used for 500ml round bottom flask. It can operate up to 650 deg.C. It features rigid housing, fabric interior and 3 in. legs.