Learn More
Chemglass Life Sciences 3000mL 3-Neck Round Bottom Flask (RBF) , 1-CN 45/50 Outer, 2-SN 24/40 Outer
SDP
Catalog No. CG152217
Encompass_Preferred
3000mL 3-Neck Round Bottomed Flask, 1-CN 45/50 Outer, 2-SN 24/40 Outer. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and vertical side necks. Flasks with 29/26 center necks have a groove tooled just below the grind to aid in clamping of the flask.
Catalog No. CG152217 Supplier Chemglass Life Sciences Supplier No. CG152217
3000mL 3-Neck Round Bottomed Flask, 1-CN 45/50 Outer, 2-SN 24/40 Outer. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and vertical side necks. Flasks with 29/26 center necks have a groove tooled just below the grind to aid in clamping of the flask.