Learn More
Chemglass Life Sciences 50mL Single Neck Round Bottom Flask (RBF) , Long Neck, 14/20 Outer Joint
SDP
Catalog No. CG151034
Encompass_Preferred
50mL Single Neck Round Bottomed Flask, Long Neck, 14/20 Outer Joint. Round bottom, HEAVY WALL single long neck flask with a standard taper outer joint.
Catalog No. CG151034 Supplier Chemglass Life Sciences Supplier No. CG151034
50mL Single Neck Round Bottomed Flask, Long Neck, 14/20 Outer Joint. Round bottom, HEAVY WALL single long neck flask with a standard taper outer joint.