Learn More
Chemglass Life Sciences 1000mL Single Neck Round Bottom Flask (RBF) , 35/25 Socket Joint, Plastic Coated
SDP
Catalog No. CG1508P34
Encompass_Preferred
1000mL Single Neck Round Bottomed Flask, 35/25 Socket Joint, Plastic Coated. Round bottom, HEAVY WALL single short neck flask for use on rotary evaporators for receiving distillate. Joint at the top of the flask is a 35/25 socket. Plastic Coated to prevent injury from broken glass while preventing spillage of hazardous materials.
Catalog No. CG1508P34 Supplier Chemglass Life Sciences Supplier No. CG1508P34
1000mL Single Neck Round Bottomed Flask, 35/25 Socket Joint, Plastic Coated. Round bottom, HEAVY WALL single short neck flask for use on rotary evaporators for receiving distillate. Joint at the top of the flask is a 35/25 socket. Plastic Coated to prevent injury from broken glass while preventing spillage of hazardous materials.