Learn More
Chemglass Life Sciences 15mL Single Neck Round Bottom Flask (RBF) , 14/20 Outer Joint
SDP
Catalog No. CG150684
Encompass_Preferred
15mL Single Neck Round Bottomed Flask, 14/20 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.
Catalog No. CG150684 Supplier Chemglass Life Sciences Supplier No. CG150684
15mL Single Neck Round Bottomed Flask, 14/20 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.