Learn More
Chemglass Life Sciences 3000mL Single Neck Round Bottom Flask (RBF) , 24/40 Outer Joint
SDP
Catalog No. CG150627
Encompass_Preferred
3000mL Single Neck Round Bottomed Flask, 24/40 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.
Catalog No. CG150627 Supplier Chemglass Life Sciences Supplier No. CG150627
3000mL Single Neck Round Bottomed Flask, 24/40 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.