Learn More
Chemglass Life Sciences 50mL Single Neck Round Bottom Flask (RBF) , 29/26 Outer Joint
SDP
Catalog No. CG150660
Encompass_Preferred
50mL Single Neck Round Bottomed Flask, 29/26 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.
Catalog No. CG150660 Supplier Chemglass Life Sciences Supplier No. CG150660
50mL Single Neck Round Bottomed Flask, 29/26 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.