Learn More
Chemglass Life Sciences 1000mL Single Neck Round Bottom Flask (RBF) , 35/20 Socket Joint
SDP
Catalog No. CG150834
Encompass_Preferred
1000mL Single Neck Round Bottomed Flask, 35/20 Socket Joint. Round bottom, HEAVY WALL single short neck flask with a spherical socket joint. Flasks having a 35/20 socket joint are used as replacements for the collection flask on rotary evaporators.
Catalog No. CG150834 Supplier Chemglass Life Sciences Supplier No. CG150834
1000mL Single Neck Round Bottomed Flask, 35/20 Socket Joint. Round bottom, HEAVY WALL single short neck flask with a spherical socket joint. Flasks having a 35/20 socket joint are used as replacements for the collection flask on rotary evaporators.