Learn More
Cole-Parmer IKA H 135.203 Flask Inlay, 50 mL, 180C max temperature
Supplier:  Cole-Parmer 0467212
Encompass
Perform multiple synthesis operations with these expansion blocks, flasks and inlays.The aluminum blocks guarantee optimal heat transfer throughout the process without interference to the magnetic field.Exchangeable flask inserts provide high-precision thermal conduction directly into the flask.
Catalog No. NC1774576