Learn More
Ace Glass, Inc. Flask, half-jacketed, three neck, 1L, 29/42 center, 24/40 sides, #11 Ace-Thred connections on inlet/outlets, for use with3/8in ID tubing
SDP
Catalog No. 50973723
Encompass
Flask, half-jacketed, three neck, 1L, 29/42 center, 24/40 sides, #11 Ace-Thred connections on inlet/outlets, for use with3/8in ID tubing
Catalog No. 50-973-723 Supplier Ace Glass, Inc. Supplier No. 6945325
Flask, half-jacketed, three neck, 1L, 29/42 center, 24/40 sides, #11 Ace-Thred connections on inlet/outlets, for use with3/8in ID tubing