Learn More
Ace Glass, Inc. Flask, half-jacketed, three neck, 3L, 34/45 center, (2) 24/40sides, 28/15 O-ring ball joint inlet/outlets
SDP
Catalog No. 50973712
Encompass
Flask, half-jacketed, three neck, 3L, 34/45 center, (2) 24/40sides, 28/15 O-ring ball joint inlet/outlets
Catalog No. 50-973-712 Supplier Ace Glass, Inc. Supplier No. 6945259
Flask, half-jacketed, three neck, 3L, 34/45 center, (2) 24/40sides, 28/15 O-ring ball joint inlet/outlets