Learn More
Ace Glass, Inc. Flask, half-jacketed, three neck, 3L, 45/50 center, (2) 24/40sides, #11 Ace-Thred connections on inlet/outlets, for use with 3/8in ID tubing
SDP
Catalog No. 50973727
Encompass
Flask, half-jacketed, three neck, 3L, 45/50 center, (2) 24/40sides, #11 Ace-Thred connections on inlet/outlets, for use with 3/8in ID tubing
Catalog No. 50-973-727 Supplier Ace Glass, Inc. Supplier No. 6945361
Flask, half-jacketed, three neck, 3L, 45/50 center, (2) 24/40sides, #11 Ace-Thred connections on inlet/outlets, for use with 3/8in ID tubing