Learn More
Ace Glass, Inc. Flask, half-jacketed, three neck, 6L, 29/42 center, (2) 29/42sides, #11 Ace-Thred connections on inlet/outlets, for use with 3/8in ID tubing
SDP
Catalog No. 50973732
Encompass
Flask, half-jacketed, three neck, 6L, 29/42 center, (2) 29/42sides, #11 Ace-Thred connections on inlet/outlets, for use with 3/8in ID tubing
Catalog No. 50-973-732 Supplier Ace Glass, Inc. Supplier No. 6945378
Flask, half-jacketed, three neck, 6L, 29/42 center, (2) 29/42sides, #11 Ace-Thred connections on inlet/outlets, for use with 3/8in ID tubing