Learn More
Ace Glass, Inc. Flask, jacketed, 2L, 100mm(4in) ground Duran; O-ring groove flange, #11 Ace-Thred™; inlet/outlet for3/8in ID tubing
SDP
Catalog No. 50973170
Encompass_Preferred
2L jktd cylindrical reaction flask with 100mm (4) Duran flat, ground flange and o-ring groove. Features a water/steam jacket with #11 Ace-Thred connections on jacket inlet/outlets for use with 3/8 I.D. tubing. Flask is supplied with one CAPFE o-ring for Duran flange.
Catalog No. 50-973-170 Supplier Ace Glass, Inc. Supplier No. 6523314
2L jktd cylindrical reaction flask with 100mm (4) Duran flat, ground flange and o-ring groove. Features a water/steam jacket with #11 Ace-Thred connections on jacket inlet/outlets for use with 3/8 I.D. tubing. Flask is supplied with one CAPFE o-ring for Duran flange.