Learn More
Ace Glass, Inc. Flask, jacketed, 15L, 200mm(8in) ground Duran; O-ring groove flange, 35/25 O-ring ball joint inlet/outlet
SDP
Catalog No. 50978795
Encompass
15L jktd cylindrical reaction flask with 200mm (8) Duran flat, ground flange and o-ring groove. Features a water/steam jacket with 35/25 o-ring ball joints on jacket inlet/outlet connections. Flask is supplied with one CAPFE o-ring for Duran flange. Flask includes reference graduations.
Catalog No. 50-978-795 Supplier Ace Glass, Inc. Supplier No. 652327
Add to Cart
Add to Cart
15L jktd cylindrical reaction flask with 200mm (8) Duran flat, ground flange and o-ring groove. Features a water/steam jacket with 35/25 o-ring ball joints on jacket inlet/outlet connections. Flask is supplied with one CAPFE o-ring for Duran flange. Flask includes reference graduations.