Learn More
Ace Glass, Inc. Flask, jacketed, 6L, 200mm(8in) ground Duran; O-ring groove flange, #15 Ace-Thred™; inlet/outlet for1/2in ID tubing
SDP
Catalog No. 50973172
Encompass_Preferred
6L jktd cylindrical reaction flask with 200mm (8) Duran flat, ground flange and o-ring groove. Features a water/steam jacket with #15 Ace-Thred connections on jacket inlet/outlets for use with 1/2 I.D. tubing. Flask is supplied with one CAPFE o-ring for Duran flange.
Catalog No. 50-973-172 Supplier Ace Glass, Inc. Supplier No. 6523319
6L jktd cylindrical reaction flask with 200mm (8) Duran flat, ground flange and o-ring groove. Features a water/steam jacket with #15 Ace-Thred connections on jacket inlet/outlets for use with 1/2 I.D. tubing. Flask is supplied with one CAPFE o-ring for Duran flange.