Learn More
Ace Glass, Inc. Flask, half-jacketed, three neck, 500mL, 24/40 center, 24/40sides, 28/15 O-ring ball joint inlet/outlets
SDP
Catalog No. 50978975
Encompass
Flask, half-jacketed, three neck, 500mL, 24/40 center, 24/40sides, 28/15 O-ring ball joint inlet/outlets
Catalog No. 50-978-975 Supplier Ace Glass, Inc. Supplier No. 6945217
Flask, half-jacketed, three neck, 500mL, 24/40 center, 24/40sides, 28/15 O-ring ball joint inlet/outlets