Learn More
Ace Glass, Inc. Flask, half-jacketed, three neck, 500mL, 29/42 center, 24/40sides, #11 Ace-Thred connections on inlet/outlets, for use with 3/8in ID tubing
SDP
Catalog No. 50978980
Encompass
Flask, half-jacketed, three neck, 500mL, 29/42 center, 24/40sides, #11 Ace-Thred connections on inlet/outlets, for use with 3/8in ID tubing
Catalog No. 50-978-980 Supplier Ace Glass, Inc. Supplier No. 6945319
Flask, half-jacketed, three neck, 500mL, 29/42 center, 24/40sides, #11 Ace-Thred connections on inlet/outlets, for use with 3/8in ID tubing