Learn More
Ace Glass, Inc. Flask, Kuderna-Danish, 1L, no hooks
SDP
Catalog No. 50978865
Encompass_Preferred
1L Kuderna-Danish flask with no hooks has a 19/22 inner joint on the bottom of the flask, and a 24/40 joint on top. For concentration of materials in volatile solvents. Combines rapidity, simplicity, and quantitative efficiency, including minimization of entrainment escape and transfer losses.
Catalog No. 50-978-865 Supplier Ace Glass, Inc. Supplier No. 670810
1L Kuderna-Danish flask with no hooks has a 19/22 inner joint on the bottom of the flask, and a 24/40 joint on top. For concentration of materials in volatile solvents. Combines rapidity, simplicity, and quantitative efficiency, including minimization of entrainment escape and transfer losses.