Learn More
Ace Glass, Inc. Flask, Kuderna-Danish, 250mL, with hooks
SDP
Catalog No. 50973377
Encompass_Preferred
250mL Kuderna-Danish flask with hooks has a 19/22 inner joint on the bottom of the flask, and a 24/40 joint on top. For concentration of materials in volatile solvents. Combines rapidity, simplicity, and quantitative efficiency, including minimization of entrainment escape and transfer losses.
Catalog No. 50-973-377 Supplier Ace Glass, Inc. Supplier No. 670811
250mL Kuderna-Danish flask with hooks has a 19/22 inner joint on the bottom of the flask, and a 24/40 joint on top. For concentration of materials in volatile solvents. Combines rapidity, simplicity, and quantitative efficiency, including minimization of entrainment escape and transfer losses.