Learn More
Ace Glass, Inc. Flask, Kuderna-Danish, 500mL, no hooks
SDP
Catalog No. 50973376
Encompass_Preferred
500mL Kuderna-Danish flask with no hooks has a 19/22 inner joint on the bottom of the flask, and a 24/40 joint on top. For concentration of materials in volatile solvents. Combines rapidity, simplicity, and quantitative efficiency, including minimization of entrainment escape and transfer losses.
Catalog No. 50-973-376 Supplier Ace Glass, Inc. Supplier No. 670809
500mL Kuderna-Danish flask with no hooks has a 19/22 inner joint on the bottom of the flask, and a 24/40 joint on top. For concentration of materials in volatile solvents. Combines rapidity, simplicity, and quantitative efficiency, including minimization of entrainment escape and transfer losses.