Learn More
Bellco Glass, Inc. Flask Locking Tray,12x12
SDP
Catalog No. 501946124
Encompass_Preferred
Includes 3 dividers. MADE IN THE USA Size: 12x12"How many size flasks will fit on the tray:125ml Erlenmeyer flasks – 16ea250ml Erlenmeyer flasks – 9ea300ml Erlenmeyer flasks – 9ea500ml Erlenmeyer flasks – 4ea1000mL Erlenmeyer flasks – 4ea2000mL Erlenmeyer flasks – 2ea
Catalog No. 50-194-6124 Supplier Bellco Glass, Inc. Supplier No. 774312012
Includes 3 dividers. MADE IN THE USA Size: 12x12"How many size flasks will fit on the tray:125ml Erlenmeyer flasks – 16ea250ml Erlenmeyer flasks – 9ea300ml Erlenmeyer flasks – 9ea500ml Erlenmeyer flasks – 4ea1000mL Erlenmeyer flasks – 4ea2000mL Erlenmeyer flasks – 2ea