Learn More
Bellco Glass, Inc. Flask Locking Tray,12x20
SDP
Catalog No. 501946125
Encompass_Preferred
Includes 5 dividers. MADE IN THE USA Size: 12x20"How many size flasks will fit on the tray:125ml Erlenmeyer flasks – 24ea250ml Erlenmeyer flasks – 15ea300ml Erlenmeyer flasks – 15ea500ml Erlenmeyer flasks – 8ea1000mL Erlenmeyer flasks – 6ea2000mL Erlenmeyer flasks – 3ea
Catalog No. 50-194-6125 Supplier Bellco Glass, Inc. Supplier No. 774312020
Includes 5 dividers. MADE IN THE USA Size: 12x20"How many size flasks will fit on the tray:125ml Erlenmeyer flasks – 24ea250ml Erlenmeyer flasks – 15ea300ml Erlenmeyer flasks – 15ea500ml Erlenmeyer flasks – 8ea1000mL Erlenmeyer flasks – 6ea2000mL Erlenmeyer flasks – 3ea