Learn More
Ace Glass, Inc. Flask, round bottom, indented morton type, three neck, 1L, 45/50center, 24/40 sides
SDP
Catalog No. 50973905
Encompass_Preferred
1L round bottom, indented, Morton-type flask with a 45/50 center neck and two angled 24/40 side necks. Indents on flask enhance agitation during stirring.
Catalog No. 50-973-905 Supplier Ace Glass, Inc. Supplier No. 695806
1L round bottom, indented, Morton-type flask with a 45/50 center neck and two angled 24/40 side necks. Indents on flask enhance agitation during stirring.