Learn More
Ace Glass, Inc. Flask, round bottom, indented morton type, three neck, 1L, 45/50center, 29/42 sides
SDP
Catalog No. 50973906
Encompass_Preferred
1L round bottom, indented, Morton-type flask with a 45/50 center neck and two angled 29/42 side necks. Indents on flask enhance agitation during stirring.
Catalog No. 50-973-906 Supplier Ace Glass, Inc. Supplier No. 695808
1L round bottom, indented, Morton-type flask with a 45/50 center neck and two angled 29/42 side necks. Indents on flask enhance agitation during stirring.