Learn More
Ace Glass, Inc. Flask, round bottom, indented morton type, three neck, 6L, 45/50center, 24/40 sides
SDP
Catalog No. 50973909
Encompass_Preferred
6L round bottom, indented, Morton-type flask with a 45/50 center neck and two vertical 24/40 side necks. Indents on flask enhance agitation during stirring.
Catalog No. 50-973-909 Supplier Ace Glass, Inc. Supplier No. 695822
6L round bottom, indented, Morton-type flask with a 45/50 center neck and two vertical 24/40 side necks. Indents on flask enhance agitation during stirring.