Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, indented morton type, three neck, 2L, 45/50 center, 29/42 sides
SDP
Catalog No. 50973914
Encompass_Preferred
2L heavy wall, round bottom, indented, Morton-type flask with a 45/50 center neck and two vertical 29/42 side necks. Indents on flask enhance agitation during stirring.
Catalog No. 50-973-914 Supplier Ace Glass, Inc. Supplier No. 6958212
2L heavy wall, round bottom, indented, Morton-type flask with a 45/50 center neck and two vertical 29/42 side necks. Indents on flask enhance agitation during stirring.