Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, indented morton type, three neck, 12L, 45/50 center, 29/42 sides
SDP
Catalog No. 5097945
Encompass_Preferred
Flask, round bottom, heavy wall, indented morton type, three neck, 12L, 45/50 center, 29/42 sides
Catalog No. 50-979-45 Supplier Ace Glass, Inc. Supplier No. 6958228
Flask, round bottom, heavy wall, indented morton type, three neck, 12L, 45/50 center, 29/42 sides