Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, indented morton type, three neck, 500mL, 45/50 center, 29/42 sides
SDP
Catalog No. 50973913
Encompass_Preferred
500mL heavy wall, round bottom, indented, Morton-type flask with a 45/50 center neck and two angled 29/42 side necks. Indents on flask enhance agitation during stirring.
Catalog No. 50-973-913 Supplier Ace Glass, Inc. Supplier No. 6958205
500mL heavy wall, round bottom, indented, Morton-type flask with a 45/50 center neck and two angled 29/42 side necks. Indents on flask enhance agitation during stirring.