Learn More
Ace Glass, Inc. Flask, nephelo, 1L, 19mm OD x 130mm sidearm, with screw caps, cs/4
SDP
Catalog No. 50971453
Encompass_Preferred
1L Nephelo culture flask with side port and angled sidearm cuvette for standard Nephelometers and spectrophotometers. Sidearm dimensions: 19mm O.D. x 130mm. Supplied with 38-460 phenolic top cap and 18-415 clean-out port cap.
Catalog No. 50-971-453 Supplier Ace Glass, Inc. Supplier No. 391035
1L Nephelo culture flask with side port and angled sidearm cuvette for standard Nephelometers and spectrophotometers. Sidearm dimensions: 19mm O.D. x 130mm. Supplied with 38-460 phenolic top cap and 18-415 clean-out port cap.